Tisk…

3-(5-Bromothiophen-2-yl)acrylic acid 95%

BLDPBD00786705-100EA 713 CZK
BLDPBD00786705-100 BLDPBD00786705-1G BLDPBD00786705-250
3-(5-Bromothiophen-2-yl)acrylic acid 95%
3-(5-Bromothiophen-2-yl)acrylic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₇H₅BrO₂S
MW: 233,0824 g/mol
Storage Temperature: Refrigerator
CAS číslo: 29079-97-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR