Tisk…

6-Bromo-5-methylquinoxaline 95%

BLDPBD00787765-250EA 6300 CZK
BLDPBD00787765-250 BLDPBD00787765-100
6-Bromo-5-methylquinoxaline 95%
6-Bromo-5-methylquinoxaline

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₉H₇BrN₂
MW: 223,0693 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31699512
CAS číslo: 2386229-26-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR