Tisk…

cis-3-(Trifluoromethyl)cyclobutan-1-amine hydrochloride 97%

BLDPBD00788334-50MEA 13630 CZK
BLDPBD00788334-50M BLDPBD00788334-100 BLDPBD00788334-250
cis-3-(Trifluoromethyl)cyclobutan-1-amine hydrochloride 97%
cis-3-(Trifluoromethyl)cyclobutan-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₅H₉ClF₃N
MW: 175,5799 g/mol
Storage Temperature: Ambient
CAS číslo: 2227198-26-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR