Tisk…

1-Methyl-1H-pyrrolo[3,2-b]pyridine-3-carboxylic acid 97%

BLDPBD00789660-250EA 3400 CZK
BLDPBD00789660-250 BLDPBD00789660-100 BLDPBD00789660-50M BLDPBD00789660-1G
1-Methyl-1H-pyrrolo[3,2-b]pyridine-3-carboxylic acid 97%
1-Methyl-1H-pyrrolo[3,2-b]pyridine-3-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₉H₈N₂O₂
MW: 176,1720 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24503553
CAS číslo: 1346608-64-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR