Tisk…

(S)-2-Amino-3-(4-((2-nitrobenzyl)oxy)phenyl)propanoic acid 98%

BLDPBD00791796-250EA 8380 CZK
BLDPBD00791796-250 BLDPBD00791796-1G BLDPBD00791796-100
(S)-2-Amino-3-(4-((2-nitrobenzyl)oxy)phenyl)propanoic acid 98%
(S)-2-Amino-3-(4-((2-nitrobenzyl)oxy)phenyl)propanoic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₁₆H₁₆N₂O₅
MW: 316,3086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31559511
CAS číslo: 184591-51-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR