Tisk…

1,1-Dimethyl-2-oxa-8-azaspiro[4.5]decane hydrochloride 97%

BLDPBD00791798-250EA 7900 CZK
BLDPBD00791798-250 BLDPBD00791798-1G BLDPBD00791798-100 BLDPBD00791798-50M
1,1-Dimethyl-2-oxa-8-azaspiro[4.5]decane hydrochloride 97%
1,1-Dimethyl-2-oxa-8-azaspiro[4.5]decane hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₀H₂₀ClNO
MW: 205,7249 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11035932
CAS číslo: 1427425-91-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR