Tisk…

2-(4-Fluorophenyl)-3-oxo-2,3-dihydropyridazine-4-carboxylic acid 95%

BLDPBD00792847-1GEA 7960 CZK
BLDPBD00792847-1G BLDPBD00792847-10G BLDPBD00792847-250 BLDPBD00792847-100 BLDPBD00792847-5G
2-(4-Fluorophenyl)-3-oxo-2,3-dihydropyridazine-4-carboxylic acid 95%
2-(4-Fluorophenyl)-3-oxo-2,3-dihydropyridazine-4-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₁H₇FN₂O₃
MW: 234,1833 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26096712
CAS číslo: 946505-09-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR