Tisk…

7-Amino-1,5-naphthyridine-3-carboxylic acid 97%

BLDPBD00793012-100EA 11550 CZK
BLDPBD00793012-100 BLDPBD00793012-250 BLDPBD00793012-1G
7-Amino-1,5-naphthyridine-3-carboxylic acid 97%
7-Amino-1,5-naphthyridine-3-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₉H₇N₃O₂
MW: 189,1708 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30828920
CAS číslo: 2089649-99-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR