Tisk…

(S)-2-((tert-Butoxycarbonyl)amino)hept-6-enoic acid 95%

BLDPBD00793303-100EA 6470 CZK
BLDPBD00793303-100
(S)-2-((tert-Butoxycarbonyl)amino)hept-6-enoic acid 95%
(S)-2-((tert-Butoxycarbonyl)amino)hept-6-enoic acid

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₂H₂₁NO₄
MW: 243,2994 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094926
CAS číslo: 204711-97-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR