Tisk…

2-Methyl-1-(piperazin-1-yl)propan-2-ol dihydrochloride 97%

BLDPBD00793640-100EA 2260 CZK
BLDPBD00793640-100 BLDPBD00793640-250 BLDPBD00793640-1G
2-Methyl-1-(piperazin-1-yl)propan-2-ol dihydrochloride 97%
2-Methyl-1-(piperazin-1-yl)propan-2-ol dihydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₂₀Cl₂N₂O
MW: 231,1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26400086
CAS číslo: 1044707-11-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR