Tisk…

2,6-Difluoro-N-(1-(4-hydroxy-2-(trifluoromethyl)benzyl)-1H-pyrazol-3-yl)benzamide 98%

BLDPBD00794291-25MEA 3800 CZK
BLDPBD00794291-25M BLDPBD00794291-250 BLDPBD00794291-50M BLDPBD00794291-100
2,6-Difluoro-N-(1-(4-hydroxy-2-(trifluoromethyl)benzyl)-1H-pyrazol-3-yl)benzamide 98%
2,6-Difluoro-N-(1-(4-hydroxy-2-(trifluoromethyl)benzyl)-1H-pyrazol-3-yl)benzamide

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₈H₁₂F₅N₃O₂
MW: 397,2988 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28411366
CAS číslo: 1253186-56-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR