Tisk…

Fmoc-4-trifluoromethyl-L-homophenylalanine 97%

BLDPBD00794882-250EA 9120 CZK
BLDPBD00794882-250 BLDPBD00794882-50M BLDPBD00794882-100
Fmoc-4-trifluoromethyl-L-homophenylalanine 97%
Fmoc-4-trifluoromethyl-L-homophenylalanine

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₂₆H₂₂F₃NO₄
MW: 469,4524 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07372279
CAS číslo: 1260591-49-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR