Tisk…

Chloromethylstyrene (m and p mixture) mixture of m- and p-isomers 88% mix stabilizovaný


Stabilization: Stabilised with TBC + ONP + o-nitrocresol

BLDPBD00795275-25GEA 884 CZK
BLDPBD00795275-25G BLDPBD00795275-5G
Chloromethylstyrene (m and p mixture) mixture of m- and p-isomers 88% mix stabilizovaný
Chloromethylstyrene (m and p mixture)

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Vzorec: C₉H₉Cl
MW: 152,62 g/mol
Storage Temperature: Freezer
MDL Number: MFCD21332905
CAS číslo: 57458-41-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR