Tisk…

Polyethylenglykol 20000 average Mn 20000

BLDPBD007962361000EA 314 CZK
BLDPBD007962361000 BLDPBD00796236-0.5 BLDPBD00796236-500
Polyethylenglykol 20000 average Mn 20000
Polyethylenglykol 20000

Storage condition: Sealed in dry, Room Temperature

Vzorec: H(OCH₂CH₂)nOH
MW: 20000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS číslo: 25322-68-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR