Tisk…

N-Fmoc-(R)-3-(methylamino)butanoic acid 98%

BLDPBD00798048-100EA 3000 CZK
BLDPBD00798048-100 BLDPBD00798048-250
N-Fmoc-(R)-3-(methylamino)butanoic acid 98%
N-Fmoc-(R)-3-(methylamino)butanoic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₂₀H₂₁NO₄
MW: 339,3851 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31580005
CAS číslo: 1460306-60-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR