Tisk…

[1-Adamantyl(phenyl)methyl]amine 97%

BLDPBD00798500-1GEA 8550 CZK
BLDPBD00798500-1G BLDPBD00798500-100 BLDPBD00798500-5G BLDPBD00798500-250
[1-Adamantyl(phenyl)methyl]amine 97%
[1-Adamantyl(phenyl)methyl]amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₇H₂₃N
MW: 241,38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02632254
CAS číslo: 139026-44-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR