Tisk…

4-Acetoxybenzyl alcohol 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD00799134-250EA 1060 CZK
BLDPBD00799134-250 BLDPBD00799134-1G BLDPBD00799134-100
4-Acetoxybenzyl alcohol 95%
4-Acetoxybenzyl alcohol

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₉H₁₀O₃
MW: 166,174 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02752162
CAS číslo: 6309-46-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR