Tisk…

4-Iodostyrene 97% stabilizovaný

Dodavatel: BLD PHARMATECH GMBH


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

BLDPBD00799731-5GEA 21690 CZK
BLDPBD00799731-5G BLDPBD00799731-1G BLDPBD00799731-250 BLDPBD00799731-100
4-Iodostyrene 97% stabilizovaný
4-Iodostyrene

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₈H₇I
MW: 230,046 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00045321
CAS číslo: 2351-50-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR