Tisk…

(6-Cyanonaphthalen-2-yl)boronic acid 95%

BLDPBD00800178-1GEA 28700 CZK
BLDPBD00800178-1G BLDPBD00800178-250 BLDPBD00800178-100
(6-Cyanonaphthalen-2-yl)boronic acid 95%
(6-Cyanonaphthalen-2-yl)boronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₁H₈BNO₂
MW: 196,9977 g/mol
Storage Temperature: Refrigerator
CAS číslo: 220299-56-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR