Tisk…

7-(Trifluoromethyl)isoquinoline 95%

BLDPBD00801509-100EA 9010 CZK
BLDPBD00801509-100 BLDPBD00801509-1G BLDPBD00801509-250
7-(Trifluoromethyl)isoquinoline 95%
7-(Trifluoromethyl)isoquinoline

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₆F₃N
MW: 197,1566 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226952
CAS číslo: 1194375-56-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR