Tisk…

6-Bromo-3-chloro-2-fluoroaniline 90%

BLDPBD00801876-100EA 1830 CZK
BLDPBD00801876-100 BLDPBD00801876-250 BLDPBD00801876-1G
6-Bromo-3-chloro-2-fluoroaniline 90%
6-Bromo-3-chloro-2-fluoroaniline
Vzorec: C₆H₄BrClFN
MW: 224,458 g/mol
MDL Number: MFCD28096616
CAS číslo: 1515343-57-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR