Tisk…

Azobenzene-4,4'-dicarbonyl dichloride 98% (by titrimetric analysis)

BLDPBD00802332-250EA 1740 CZK
BLDPBD00802332-250 BLDPBD00802332-100 BLDPBD00802332-1G BLDPBD00802332-5G
Azobenzene-4,4'-dicarbonyl dichloride 98% (by titrimetric analysis)
Azobenzene-4,4'-dicarbonyl dichloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₄H₈Cl₂N₂O₂
MW: 307,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053223
CAS číslo: 10252-29-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR