Tisk…

(S)-Fmoc-2-amino-3-tert-butoxy-2-methyl-propionic acid 97%

BLDPBD00802618-100EA 1540 CZK
BLDPBD00802618-100 BLDPBD00802618-1G BLDPBD00802618-250
(S)-Fmoc-2-amino-3-tert-butoxy-2-methyl-propionic acid 97%
(S)-Fmoc-2-amino-3-tert-butoxy-2-methyl-propionic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₂₃H₂₇NO₅
MW: 397,4642 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26793609
CAS číslo: 914399-98-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR