Tisk…

2-(Undecyloxy)ethanol 98%

BLDPBD00804095-250EA 684 CZK
BLDPBD00804095-250 BLDPBD00804095-1G BLDPBD00804095-25G BLDPBD00804095-5G
2-(Undecyloxy)ethanol 98%
2-(Undecyloxy)ethanol

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₃H₂₈O₂
MW: 216,3700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06252492
CAS číslo: 38471-47-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR