Tisk…

1,2-distearoyl-sn-glycerol 97%

BLDPBD00804304-250EA 2110 CZK
BLDPBD00804304-250 BLDPBD00804304-100 BLDPBD00804304-1G
1,2-distearoyl-sn-glycerol 97%
1,2-distearoyl-sn-glycerol

Storage condition: Sealed in dry, Store in freezer, under -20°C

Vzorec: C₃₉H₇₆O₅
MW: 625,032 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00067473
CAS číslo: 10567-21-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR