Tisk…

7-Bromo-5-fluoro-3,4-dihydroisoquinolin-1(2H)-one 95%

BLDPBD00805010-1GEA 30470 CZK
BLDPBD00805010-1G BLDPBD00805010-100 BLDPBD00805010-250
7-Bromo-5-fluoro-3,4-dihydroisoquinolin-1(2H)-one 95%
7-Bromo-5-fluoro-3,4-dihydroisoquinolin-1(2H)-one

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₇BrFNO
MW: 244,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20287127
CAS číslo: 1341674-79-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR