Tisk…

2-Methoxypyridine-4-sulphonyl chloride 98%

BLDPBD00807182-250EA 4250 CZK
BLDPBD00807182-250 BLDPBD00807182-100 BLDPBD00807182-1G
2-Methoxypyridine-4-sulphonyl chloride 98%
2-Methoxypyridine-4-sulphonyl chloride

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₆ClNO₃S
MW: 207,6347 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13188796
CAS číslo: 1060807-43-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR