Tisk…

(3S)-4,4-Dimethyl-pyrrolidine-3-carboxylic acid 96%

BLDPBD00807431-100EA 9950 CZK
BLDPBD00807431-100 BLDPBD00807431-250 BLDPBD00807431-25M BLDPBD00807431-50M
(3S)-4,4-Dimethyl-pyrrolidine-3-carboxylic acid 96%
(3S)-4,4-Dimethyl-pyrrolidine-3-carboxylic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₇H₁₃NO₂
MW: 143,1836 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201190
CAS číslo: 1373232-20-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR