Tisk…

(S)-1-Boc-3-(aminocarbonyl)piperidine 95%

BLDPBD00807606-250EA 1200 CZK
BLDPBD00807606-250 BLDPBD00807606-5G BLDPBD00807606-1G
(S)-1-Boc-3-(aminocarbonyl)piperidine 95%
(S)-1-Boc-3-(aminocarbonyl)piperidine

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₁H₂₀N₂O₃
MW: 228,2881 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977786
CAS číslo: 88466-77-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR