Tisk…

1-Boc-D-prolyl-glycine

Dodavatel: BLD pharmatech
BLDPBD00807734-1GEA 5450 CZK
BLDPBD00807734-1G BLDPBD00807734-5G BLDPBD00807734-250
1-Boc-D-prolyl-glycine
Proteiny
  • GHS07

Storage condition: Sealed in dry, 2-8°C
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR