Tisk…

2-(2-Bromophenyl)-9-phenyl-9H-carbazole 98%

BLDPBD00807752-100EA 912 CZK
BLDPBD00807752-100
2-(2-Bromophenyl)-9-phenyl-9H-carbazole 98%
2-(2-Bromophenyl)-9-phenyl-9H-carbazole

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₂₄H₁₆BrN
MW: 398,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27644694
CAS číslo: 1616607-88-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR