Tisk…

2-(5,6-Difluoro-3-oxo-2,3-dihydro-1H-inden-1-ylidene)malononitrile 98%

BLDPBD00807938-250EA 2460 CZK
BLDPBD00807938-250 BLDPBD00807938-5G BLDPBD00807938-1G
2-(5,6-Difluoro-3-oxo-2,3-dihydro-1H-inden-1-ylidene)malononitrile 98%
2-(5,6-Difluoro-3-oxo-2,3-dihydro-1H-inden-1-ylidene)malononitrile

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₂H₄F₂N₂O
MW: 230,1698 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31631729
CAS číslo: 2083617-82-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR