Tisk…

4,5-Difluoro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine 98%

BLDPBD00808084-100EA 10780 CZK
BLDPBD00808084-100 BLDPBD00808084-1G BLDPBD00808084-250
4,5-Difluoro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine 98%
4,5-Difluoro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Vzorec: C₁₆H₂₄F₂N₂Si
MW: 310,4575 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11616415
CAS číslo: 685513-89-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR