Tisk…

4-(2,2-Diethoxyethoxy)but-1-ene 90% stabilizovaný


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

BLDPBD00808723-250EA 5080 CZK
BLDPBD00808723-250 BLDPBD00808723-1G BLDPBD00808723-100
4-(2,2-Diethoxyethoxy)but-1-ene 90% stabilizovaný
4-(2,2-Diethoxyethoxy)but-1-ene

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₀H₂₀O₃
MW: 188,2640 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18848577
CAS číslo: 1343915-25-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR