Tisk…

6-Bromo-2,8-dimethyl-[1,2,4]triazolo[1,5-a]pyridine 95%

BLDPBD00808808-1GEA 9120 CZK
BLDPBD00808808-1G BLDPBD00808808-100 BLDPBD00808808-250
6-Bromo-2,8-dimethyl-[1,2,4]triazolo[1,5-a]pyridine 95%
6-Bromo-2,8-dimethyl-[1,2,4]triazolo[1,5-a]pyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₈H₈BrN₃
MW: 226,0732 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26732010
CAS číslo: 1554779-20-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR