Tisk…

Iron(III) stearate 98%

BLDPBD00809031-100EA 1460 CZK
BLDPBD00809031-100 BLDPBD00809031-25G BLDPBD00809031-5G
Iron(III) stearate 98%
Iron(III) stearate

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₈H₃₆O₂
MW: 906,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050716
CAS číslo: 555-36-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR