Tisk…

Methyl (2R)-2-hydroxy-4-methylpentanoate 95%

BLDPBD00810348-100EA 884 CZK
BLDPBD00810348-100 BLDPBD00810348-1G BLDPBD00810348-250 BLDPBD00810348-5G
Methyl (2R)-2-hydroxy-4-methylpentanoate 95%
Methyl (2R)-2-hydroxy-4-methylpentanoate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₁₄O₃
MW: 146,1843 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20482702
CAS číslo: 112529-90-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR