Tisk…

Polyethylene glycol monomethyl ether, M.W. ~4000

BLDPBD00812118-25GEA 485 CZK
BLDPBD00812118-25G BLDPBD00812118-500 BLDPBD00812118-100
Polyethylene glycol monomethyl ether, M.W. ~4000
Polyethylene glycol monomethyl ether

Storage condition: Sealed in dry, Room Temperature

Vzorec: CH₃(OCH₂CH₂)nOH
Storage Temperature: Ambient
MDL Number: MFCD00084416
CAS číslo: 9004-74-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR