Tisk…

(4,4-Difluorocyclohexyl)hydrazine hydrochloride 97%

BLDPBD00812897-100EA 3400 CZK
BLDPBD00812897-100 BLDPBD00812897-1G BLDPBD00812897-250
(4,4-Difluorocyclohexyl)hydrazine hydrochloride 97%
(4,4-Difluorocyclohexyl)hydrazine hydrochloride

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₆H₁₃ClF₂N₂
MW: 186,6306 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30802276
CAS číslo: 1548590-10-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR