Tisk…

7-Phenyl-1,2,3,4-tetrahydroquinoline 95%

BLDPBD00813596-1GEA 10490 CZK
BLDPBD00813596-1G BLDPBD00813596-250 BLDPBD00813596-100
7-Phenyl-1,2,3,4-tetrahydroquinoline 95%
7-Phenyl-1,2,3,4-tetrahydroquinoline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₅H₁₅N
MW: 209,2900 g/mol
Storage Temperature: Ambient
CAS číslo: 60640-17-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR