Tisk…

6-Iodo-8-phenylimidazo[1,2-a]pyrazine 95%

BLDPBD00816835-250EA 19410 CZK
BLDPBD00816835-250 BLDPBD00816835-100
6-Iodo-8-phenylimidazo[1,2-a]pyrazine 95%
6-Iodo-8-phenylimidazo[1,2-a]pyrazine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₁₂H₈IN₃
MW: 321,116 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31561255
CAS číslo: 2227272-86-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR