Tisk…

Ethyl 1-(4-chlorophenyl)cyclopropanecarboxylate 97%

BLDPBD00816982-1GEA 10320 CZK
BLDPBD00816982-1G BLDPBD00816982-250 BLDPBD00816982-100
Ethyl 1-(4-chlorophenyl)cyclopropanecarboxylate 97%
Ethyl 1-(4-chlorophenyl)cyclopropanecarboxylate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₂H₁₃ClO₂
MW: 224,6800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09751950
CAS číslo: 148992-58-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR