Tisk…

5-Bromo-2-methoxy-6-methylpyridine-3-carboxylic acid 95%

BLDPBD00817525-1GEA 25260 CZK
BLDPBD00817525-1G BLDPBD00817525-100 BLDPBD00817525-250
5-Bromo-2-methoxy-6-methylpyridine-3-carboxylic acid 95%
5-Bromo-2-methoxy-6-methylpyridine-3-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₈BrNO₃
MW: 246,0580 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22033941
CAS číslo: 1369354-33-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR