Tisk…

N-Cyclohexylacetamide 95%

BLDPBD00820591-100EA 1970 CZK
BLDPBD00820591-100 BLDPBD00820591-1G BLDPBD00820591-250
N-Cyclohexylacetamide 95%
N-Cyclohexylacetamide

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₁₅NO
MW: 141,2108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00982151
CAS číslo: 1124-53-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR