Tisk…

(3-Chloro-6-fluoro-2-methoxyphenyl)boronic acid 95%

BLDPBD00820772-250EA 1740 CZK
BLDPBD00820772-250 BLDPBD00820772-1G BLDPBD00820772-100
(3-Chloro-6-fluoro-2-methoxyphenyl)boronic acid 95%
(3-Chloro-6-fluoro-2-methoxyphenyl)boronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₇BClFO₃
MW: 204,3911 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22414969
CAS číslo: 2121511-91-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR