Tisk…

N-(3-Bromo-4-methylphenyl)-4-(pyridin-4-ylmethyl)phthalazin-1-amine 98%

BLDPBD00821487-50MEA 3170 CZK
BLDPBD00821487-50M BLDPBD00821487-5MG BLDPBD00821487-10M BLDPBD00821487-1MG BLDPBD00821487-25M
N-(3-Bromo-4-methylphenyl)-4-(pyridin-4-ylmethyl)phthalazin-1-amine 98%
N-(3-Bromo-4-methylphenyl)-4-(pyridin-4-ylmethyl)phthalazin-1-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₂₁H₁₇BrN₄
MW: 405,2905 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17014834
CAS číslo: 300842-64-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR