Tisk…

6-Fluoro-1H-indazol-7-amine 97%

BLDPBD00822867-1GEA 13400 CZK
BLDPBD00822867-1G BLDPBD00822867-100 BLDPBD00822867-250
6-Fluoro-1H-indazol-7-amine 97%
6-Fluoro-1H-indazol-7-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₇H₆FN₃
MW: 151,1400 g/mol
Storage Temperature: Ambient
CAS číslo: 1780425-08-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR