Tisk…

1,3-Dibromo-5-butoxybenzene 95%

BLDPBD00823174-250EA 2340 CZK
BLDPBD00823174-250 BLDPBD00823174-1G BLDPBD00823174-5G
1,3-Dibromo-5-butoxybenzene 95%
1,3-Dibromo-5-butoxybenzene

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₁₂Br₂O
MW: 308,0097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043831
CAS číslo: 896745-35-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR