Tisk…

2-(2-(2-((Tetrahydro-2H-pyran-2-yl)oxy)ethoxy)ethoxy)ethan-1-ol 95%

BLDPBD00824204-1GEA 1830 CZK
BLDPBD00824204-1G BLDPBD00824204-25G BLDPBD00824204-5G
2-(2-(2-((Tetrahydro-2H-pyran-2-yl)oxy)ethoxy)ethoxy)ethan-1-ol 95%
2-(2-(2-((Tetrahydro-2H-pyran-2-yl)oxy)ethoxy)ethoxy)ethan-1-ol

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₁H₂₂O₅
MW: 234,2894 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28976687
CAS číslo: 60221-37-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR