Tisk…

N-(Adamantan-1-yl)quinoxaline-2-carboxamide 98%

BLDPBD00824272-250EA 11830 CZK
BLDPBD00824272-250 BLDPBD00824272-25M BLDPBD00824272-100 BLDPBD00824272-50M
N-(Adamantan-1-yl)quinoxaline-2-carboxamide 98%
N-(Adamantan-1-yl)quinoxaline-2-carboxamide

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₉H₂₁N₃O
MW: 307,3895 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865239
CAS číslo: 226878-01-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR